Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ alpha Satellite Repeat Primer
![SDP](/content/dam/fishersci/glyphs/sdp.png)
alpha SAT primer for PCR in chromatin precipitation
Supplier: Novus Biologicals™ NBP171654
Description
- Primer sets come premixed with forward and reverse primers at a concentration of 10μM
- Primers are compatible with all real-time PCR chemistries
- Each primer set has been tested and validated by chromatin immunoprecipitation (ChIP)
- Tm for primer set is between 53.4° to 54°c
- Amplicon size of primer set is 150 base pairs
Recommended Storage
Store at -20°C. Avoid freeze-thaw cycles.
Specifications
alpha Satellite Repeat Primer | |
Store at –20°C. Avoid Free/Thaw Cycles | |
0.5 mL | |
Chromatin Immunoprecipitation |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction