Learn More
Novus Biologicals™ AbSelect™ Generic Antibody Purification Kit
Fast and simple method to purify antibodies from ascites fluid and serum
Supplier: Novus Biologicals™ 8600005
Description
The AbSelect method involves capture of the antibody on protein A resin and the removal of unwanted substances by a simple wash procedure. The purified product is then eluted and neutralized. The antibody to be purified or cleaned up ideally is in a volume of 100μL to 0.5mL, up to 500μg of antibody can be purified in each run.
Compatible with lightning-link antibody labeling system, which allows purified antibody to be rapidly conjugated with wide variety of labelsRecommended Storage
Store at 4°C. Do not freeze.
Specifications
Antibody Purification Kit | |
1 Purification | |
4°C. Do not freeze. | |
1 Test |