Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Novagen™ T7·Tag™ Monoclonal Antibody
Labeled with biotin, direct conjugate with alkaline phosphatase or horseradish peroxidase.
$324.00 - $1271.00
Description
- Directed against 11 amino acid gene 10 leader peptide expressed by many of the pET translation vectors
- Cross-reactivity: Negligible with bacterial, insect or mammalian cell lysates
- Sensitivity 1-10ng (Western blot developed with chromogenic substrates)
- Suitable for immunoprecipitation immunohistochemistry (1-3)
Specifications
-20°C | |
T7 Monoclonal Antibody |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title