Learn More
MilliporeSigma™ Novagen™ λDE3 Lysogenization Kit
Construction of bacterial strains that carry inducible T7 RNA Polymerase. MilliporeSigma™; Novagen™ λDE3 Lysogenization Kit is designed for site-specific integration of λDE3 prophage into an E. coli host cell chromosome.
Supplier: MilliporeSigma™ 697343
Includes:
200μL λDE3 Phage Lysate, 200μL Helper Phage Lysate, 200μL Selection Phage Lysate, 50μL T7 Tester Phage Lysate, 200μL HMS174(DE3) glycerol stock
Description
- Used to express target genes cloned in pET vectors
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
10 Reactions | |
λDE3 Lysogenization Kit | |
Store at ≤70°C. Avoid freeze/thaw. Ok to freeze. |