Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Notrax™ 194CW Keep Calm and Wash Hands Mat
Safety Message mats make safety message loud and clear while keeping facilities cleaner and safer.
$227.50 - $373.50
Specifications
Color | Blue/White |
---|---|
Material | Nylon, Vinyl |
Type | Entrance Mat |
Thickness (English) | 0.38 in. |
Thickness (Metric) | 0.97 cm |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-085-909
|
NoTrax
194SCW35BU |
5 x 3 ft. (152 x 91 cm) |
Each for $227.50
|
|
|||||
19-085-910
|
NoTrax
194SCW46BU |
6 x 4 ft. (183 x 122 cm) |
Each for $373.50
|
|
|||||
Description
- Pre-printed message mats warn employees or customers who may be entering a hazardous area
- Offer functionality as an entrance mat, cleaning dirt and moisture from shoes while keeping facilities cleaner and safer.
- Safety Message Mats should be used indoors at entrances, lobbies, foyers, or any other light to heavy traffic areas where a safety message should be seen.
- Highly visible colors and graphics allow for immediate identification
- 24 ounce nylon top surface provides excellent moisture absorption and retention
- Vinyl backing ensure minimal buckling and mat movement
Specifications
Blue/White | |
Entrance Mat | |
0.97 cm |
Nylon, Vinyl | |
0.38 in. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title