Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Redi-Care First Aid Kits

Lightweight and easy to tote

Supplier:  Honeywell™ 0185004222

 View more versions of this product

Catalog No. 19-014-835

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.


Description

Description

Lightweight and easy to tote

Portable general-industry first aid kits are available in several sizes and packages. Kits' contents come in lightweight, soft-sided nylon bags with carrying handles and belt loops, making them easy to tote. High visibility red.

  • Promotional/Individual kit includes 37 items, and measures 1.50L × 4.5W × 7 in.H (3.81 × 11.43 × 17.78cm)
  • Small kit includes 42 items and measures 6.4 × 12.7 × 12.1 in. (16.3 × 32.3 × 30.7cm)
  • Medium kit includes 133 items, and measures 8.75L × 6W × 2.75 in.H (22.2 × 15.2 × 7.0cm)
  • Large kit includes 248 items and measures 10.5L × 7W × 6 in.H (26.7 × 17.8 × 15.2cm)

Order Info

Medium and large kits are also available with a CPR barrier.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Redi-Care First Aid Kit
Used in oil and gas industries, pharmaceutical industries, municipal services
10.5 in.
Red
Large
8 in.
15.24 cm
6 in.
26.67 cm
ANSI Z308.1-2003
Hundred 1 — 3 in. cloth adhesive bandages, four cloth adhesive bandages extra large, four cloth adhesive bandages fingertip, six cloth adhesive bandages knuckle, 0.5 in. — 2.5 yd. Adhesive tape, 1 in. — 5 yd. Adhesive tape, ten Antibiotic ointment pouch,
10.5 x 8 x 6 in. (26.67 x 25.4 x 15.24 cm)
25.4 cm
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit