Learn More
Honeywell™ North™ Protecto-Shield Visor Headgear
Tough faceshield protection with PresLock or Ratchet suspension adjustable headgear. Fits most commonly used visors. This complete faceshield is a V5N Protecto-Shield ProLok headgear with clear polycarbonate visor.
Supplier: Honeywell™ 11340145
Description
- Constructed of tough thermoplastic for superior heat protection
- Designed with a chemical splash drip edge to prevent fluids from splashing down inside the visor
- Easy sizing and tension adjustment for individual visor positioning
- Visors easily attach with no snaps, posts or levers
- Available with ratchet headband with tension knob that allows easy sizing and adjustment for individual visor positioning
- Available with pinlock headband that provides adjustments similar to a baseball cap
- Available in propionate, polycarbonate and acetate for a variety of environments
- Popular industry sizes and thicknesses
- Easy attachment without snaps, posts or levers; reinforced edge to resist cracking
- Universal hole pattern allows the use of Sperian visors with other popular manufacturers' headgear
- Aluminum bound acetate replacement visors form easily for a contoured, customized fit