Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ North™ Plastic Tweezers
Honeywell™ North™ Plastic Tweezers with pointed blade ends and corrugate handle for comfort grip.
Supplier: Honeywell™ 031570K
This item is not returnable.
View return policy
Description
- Black Forceps made of plastic for long durability
- Forceps with pointed blade ends and corrugate handle for comfort grip
Specifications
First Aid Products | |
33.43 cm | |
Black | |
1 Ea. |
Wound Care | |
4 in. | |
Plastic | |
Pointed |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction