Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Faceshield Window

Catalog Number 19823345
Click to view available options
Product Type:
Faceshield window, Shade 5
Faceshield window, Shade 8
This item is not returnable. View return policy
This item is not returnable. View return policy
Width (Metric) 41.9 cm
Thickness (English) 0.060 in.
Width (English) 16.5 in.
Product Type Faceshield window, Shade 8
Length (English) 8 in.
Length (Metric) 20.3 cm
Lens Material Fiber-metal
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit