Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ North™ Primair™ 100 Series PA101 Loose-Fitting Facepiece Assembly
Loose-fitting assemblies are designed for pharmaceutical and other light industries.
Supplier: Honeywell™ PA101S
Includes: Facepiece, adjustable headgear, and replaceable sweatband.
Description
- Spun-bound polypropylene hood cover with Velcro™ strip and rust-resistant snaps attaches and removes easily from headgear
- Triple adjustment headgear provides a secure and comfortable fit
- Replaceable 3/16 in. (0.48 cm) foam sweatband with adhesive backing adheres and removes easily from headband
- Lens made of 0.040 mil co-polyester
- Polypropylene chin cover
- Hoods with collar and full bib have an assigned protection factor (APF) of 1000
- Manifold directs air over lens to reduce fogging
Specifications
25 | |
Polypropylene | |
Small | |
Pharmaceutical and light duty applications | |
Tychem™ QC headcover and adjustable headgear | |
PA101S |
NIOSH | |
Facepiece Assembly | |
Continuous flow | |
Compact Air 200 Series PAPR and CF1000 Series | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction