Learn More
Vector Laboratories Normal Rabbit Serum Blocking Solution 20 ml
Supplier: Vector Laboratories S500020
This product is made from sera collected from healthy adult rabbits. Serum pooled from several animals is heat-treated, centrifuged to remove precipitates and filtered. The sera are tested with the appropriate antibody to check for possible cross-reactivities and supplied undiluted with 0.08% sodium azide as a preservative. When diluted, this reagent can be used for blocking non-specific binding when rabbit secondary antibodies are used in immunohistochemistry (IHC) assays. It can also be used as an antibody diluent.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.