Learn More
Immunology Consultants Normal Rabbit Serum, 100 ml
Supplier: Immunology Consultants RS15100ML
Normal rabbit serum was collected from healthy rabbits and sterile filtered. Optimal working dilutions should be determined experimentally by the investigator.