Learn More
Utak Normal Human Serum, 500mL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Utak 44600S(500)
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
100% Real Normal Human Serum is collected in FDA-licensed facilities within the United States from healthy adult donors. The Normal Human Serum is screened for common drugs of abuse and alcohol. Frozen serum is suitable for all testing methods, including LCMS, LC-MS/MS, GCMS, Time of Flight (TOF), ELISA, EMIT and other immunoassays. This product uses Potassium Oxalate as an anticoagulant and Sodium Fluoride for preservation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.