Learn More
Quidel Corporation Normal Human Serum Standard Complement 2.5ml
Supplier: Quidel Corporation A112
Uniform pool of human serum complement that has been characterized for levels of complement activation fragments as well as CH50.