Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Drierite™ Drying Desiccants, W.A. Hammond

Click to view available options
Quantity:
1 lb.
5 lb.
25 lb.
Mesh Size:
Nonindicating; 10-20 Mesh; 1 lb.
Nonindicating; 10-20 Mesh; 5 lb.
Nonindicating; 4 Mesh; 5 lb.
Nonindicating; 4 mesh; 1lb
Nonindicating; 6 Mesh, 1 lb.
Nonindicating; 6 Mesh; 5 lb.
Nonindicating; 8 Mesh; 1 lb.
Nonindicating; 8 Mesh; 5 lb.
Nonindicating; 8 mesh; 25 lb.
Description
Dries air and gases to lowest residual moisture level. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.
- All purpose desiccant made of anhydrous calcium sulfate
- Dries air and gases to the lowest residual moisture level attainable with a safe, easy to use desiccant (-100°F dew point)
- Dries organic liquids to low ppm levels
- Commonly used in desiccators, desiccator cabinets, in-line air and gas dryers, glove boxes, sealed chambers
- Useful for water removal from liquid reaction mixtures and multi-component gas streams.
- Neutral, stable, constant in volume, and inert except toward water
- Insoluble in organic solvents
- Absorbs moisture at temperatures up to 66°C
Specifications
CAS | 7778-18-9 |
Material | Calcium Sulfate |
Quantity | 25 lb. |
Mesh Size | Nonindicating; 8 mesh; 25 lb. |
Packaging | Glass Bottle |
Synonym | Abosrbent |
Chemical Name or Material | Calcium Sulfate |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title