Learn More
Fine Science Tools Scalpel Blades - #10, 100/pk
Supplier: Fine Science Tools 1001000
Product: Scalpel Blades - #10/ Blade Shape: Curved/ Cutting Edge: 30mm/ Thickness: 0.4mm/ Alloy / Material: Carbon Steel. Blades come individually wrapped in transparent packages. The alloy has a high carbon content which provides a longer lasting cutting edge. Not autoclavable. 100/pk