Learn More
Chemglass Life Sciences #15 Internal Thread, Glass Only
Supplier: Chemglass Life Sciences CG36003
#15 Internal Thread, Glass Only. Internal glass thread for use with CG-362, CG-363, CG-364, and CG-366 PTFE bushings. Tubing OD listed is the shank OD of the thread.Internal Thread Specifications:#7 Internal Thread: 12.7mm Tube OD, Accepts 6-7mm OD Tubing#11 Internal Thread: 16mm Tube OD, Accepts 9-10.5mm OD Tubing#15 Internal Thread: 22mm Tube OD, Accepts 12.5-14mm OD Tubing#25 Internal Thread: 32mm Tube OD, Accepts 24-25mm OD Tubing#50 Internal Thread: 57mm Tube OD, Accepts 47-48mm OD Tubing
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.