Learn More
Wilmad Labglass NMR TUBE PREC 5MMX7 300MHZ 5PK
Supplier: Wilmad Labglass 507PP75
5 Mm Thin Wall Precision Nmr Sample Tube 7" L, 300Mhz Wilmad-Labglass'S Nmr Precision Tubes Are Manufactured From Borosilicate Glass That Meets The Requirement Of Type 1 Class A Glass From Astm E438. Precision Tube Has 3 Fold Less Paramagnetic Contamination Comparing To The Economy Tube, And Can Be Operated Safely At Temperature Up To 230 ˚C, And Within A Temperature Step Of 120 ˚C. Precision Tube Is Rated As Hydrolytic Class 1, Which States That The Inner Surface Is Resistant To Strong Acid And Base At Ambient Temperature.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.