Learn More
Chemglass Life Sciences NMR Tube Cleaner, Complete
Supplier: Chemglass Life Sciences CG185401
NMR Tube Cleaner, Complete. All glass and PTFE design cleans five NMR tubes of the same, or various, lengths and diameters. PTFE adapter has five positions with flexible 1/8” PTFE tubing supplied in 9” lengths that can be cut to desired height. The NMR tube is placed over the tubing and seats in the adapter, leaving a small space between the end of the PTFE tubing and the inside bottom of the NMR tube. With the stopcock in the closed position, the reservoir is connected to a low vacuum source. Cleaning solvent is added into the adapter followed by air drying to complete the process in seconds. Flexible tubing and adapter design reduces breakage. Stopcock permits easy draining of solvent. Hose connection is 10mm at the largest serration.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.