Learn More
Chemglass Life Sciences Assortment Box: NMR Tube Caps, 100 of each CG1842 cap color
Supplier: Chemglass Life Sciences CG184210
Assortment Box: NMR Tube Caps, 100 of each CG1842 cap color (black, white, yellow, red, green, and blue.)Made from polyethylene, these caps are sized to fit 5mm O.D. NMR sample tubes. Cap fits tightly to stay in place during increased internal vaporization pressure. Available in a pack of 100, or in a convenient divided polypropylene box having 100 each of the 6 listed colors.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.