Learn More
High Purity Standards NITRITE STD 1000PPM 250ML
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: High Purity Standards ICNM250
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
1000 ug/mL Nitrite in H2O; Single Component IC standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.