Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety NitriShield™ with Grippaz™ Technology, Orange Nitrile Gloves

Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

The unique fish-scale pattern of these orange nitrile gloves provides unsurpassed grip and durability. Ergonomically designed for wet grip on the outside and to reduce hand slippage on the inside.

  • 6 mil, Orange Powder Free Nitrile
  • Fish-scale pattern grip
  • Textured inside for better traction
  • 9.5 Inch Industrial/food service grade
  • Component materials comply with all relevant federal regulations for food contact
Certifications/Compliance FDA Compliant
Applications Industrial
AQL 1.5
Color Orange
Disposable Yes
Exterior Finish Fully Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size 2X-Large
Thickness (English) 6 mil
Thickness (Metric) 0.15 mm
Model 6016OXXL
Show More Show Less