Learn More
Bioassay Systems EnzyChrom™ Nitric Oxide Synthase Assay Kit. For quantitative determination of nitric oxide synthase (NOS) activity and screen for NOS inhibitor.
Supplier: Bioassay Systems ENOS100
For quantitative determination of nitric oxide synthase (NOS) activity and screen for NOS inhibitor. Key Features: Sensitive and accurate. Detection range 0.25 - 25 U/L in 96-well plate. Rapid and reliable. Can be completed in 40 min if reduction of NO3- to NO2- is performed at 60°C. Method: OD540nm. Samples: Biological. Species: All. Procedure: Assay takes 40 min. Kit size: 100 tests. Detection limit: 0.25 U/L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.