Learn More
Nitinol foil, 0.38mm (0.015in) thick, superelastic, flat annealed,pickled surface, Thermo Scientific Chemicals
Form: Approx.1.56g/25 x 25mm; Af Approx. 10deg.C; Superelastic; Pickled surface
$69.65 - $226.65
Specifications
Chemical Name or Material | Nitinol |
---|---|
Color | Silver to Gray |
Name Note | Flat annealed |
Appearance | Silver-gray foil |
MDL Number | MFCD02091734 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA44954FF
|
Thermo Scientific Chemicals
044954FF |
25 x 25 mm |
Each
for $69.65
Save 13% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA44954FI
|
Thermo Scientific Chemicals
044954FI |
50 x 50 mm |
Each
for $226.65
Save 12% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Nitinol foil is used in photo chemically-etched parts, formed sheet shapes, EDM or laser cut shapes, cell-phone technology as a retractable antenna, or microphone boom.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nitinol | |
Flat annealed | |
MFCD02091734 | |
Odorless |
Silver to Gray | |
Silver-gray foil | |
Insoluble in water. | |
Foil |
Safety and Handling
Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.
H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.