Learn More
Nitinol foil, 0.127mm (0.005in) thick, superelastic, flat annealed, pickled surface, Thermo Scientific Chemicals
$78.65 - $255.65
Specifications
Chemical Name or Material | Nitinol foil |
---|---|
Name Note | 0.127mm (0.005 in.) thick, superelastic, flat annealed, pickled surface |
MDL Number | MFCD02091734 |
Solubility Information | Insoluble in water. |
Odor | Odorless |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA44952FF
|
Thermo Scientific Chemicals
044952FF |
25 x 25 mm |
Each
for $78.65
Save 16% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA44952FI
|
Thermo Scientific Chemicals
044952FI |
50 x 50 mm |
Each
for $255.65
Save 16% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Innovations in the manufacturing and processing of Nitinol have led to a great expansion of Nitinol medical devices in a variety of therapeutic areas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nitinol foil | |
MFCD02091734 | |
Odorless |
0.127mm (0.005 in.) thick, superelastic, flat annealed, pickled surface | |
Insoluble in water. | |
≈0.51g/25x25mm |
Safety and Handling
GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.