Learn More
Beckman Coulter NIST Traceable Particle Size Standards
For calibrating Coulter Counter Aperture Tubes, and verifying their performance.
Supplier: Beckman Coulter 6601329
Description
- Use the recommended bead with primary aperture for Aperture Calibration
- Additional sized beads can be used to verify aperture performance after calibration
- L10 and L20 standards are included with the Multisizer 4e Coulter Counter Sales Group
- NIST Traceable
Specifications
Particle Size Standard | |
Latex | |
Calibration and Performance Verification |
5 x 15 mL | |
2 μm, 5 μm, 10 μm, 20 μm, 43 μm | |
Aperature Tubes |