Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beckman Coulter NIST Traceable Particle Size Standards
For calibrating Coulter Counter Aperture Tubes, and verifying their performance.
Supplier: Beckman Coulter 6602800
Description
- Use the recommended bead with primary aperture for Aperture Calibration
- Additional sized beads can be used to verify aperture performance after calibration
- L10 and L20 standards are included with the Multisizer 4e Coulter Counter Sales Group
- NIST Traceable
Specifications
Particle Size Standard | |
Polystyrene | |
Calibration and Performance Verification |
15 mL | |
43 μm | |
Aperature Tubes (280 μm) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title