Learn More
Immunochemistry Technology Nigericin 0.5 µmoles, 25 -50 test size
Supplier: Immunochemistry Technology 6698
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Nigericin is a potent microbial toxin that acts as a potassium ionophore, inducing a net decrease in intracellular levels of potassium. This is critical for the oligomerization of the NLRP3 inflammasome and activation of caspase-1 in pyroptosis. Nigericin can be used as a positive control in pyroptosis experiments.