Learn More
Perkin Elmer US LLC Nickel Sampler Cone for NexION 300/350/1000/2000/5000
Supplier:  Perkin Elmer US LLC W1033612
Encompass_Preferred
Precision-designed and manufactured, large-orifice sampler and skimmer cones provide superior long-term stability and resist clogging, allowing analysis under both high and low sample-uptake conditions. Nickel is a very rugged, long-lasting material for the majority of sample types, while platinum is the material of choice for more corrosive samples. They have been designed to maximize signal stability and to minimize clogging during extended runs of samples containing high dissolved solids.
Catalog No. NC0050285