Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nickel-Plated Forceps



Handle dissections economically.
Supplier: Humboldt Manufacturing Company H12250
Description
Affordable, straight-type steel forceps will easily handle dissection duty.
- Dimensions: 5 in. (12.5cm)
Specifications
Corrugated, Straight | |
Steel, Nickel Plated | |
Forceps |
5 in. | |
127 mm | |
Each |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title