Learn More
Roche Diagnostics NH3 Bio HT, Test kit for the Cedex Bio HT Analyzer
Supplier: Roche Diagnostics 06608515001
Test kit for the Cedex Bio HT analyzer. Kit for determination of ammonia. Enzymatic amination by glutamate dehydrogenase; recovery a+/- 10%
This item is not returnable.
View return policy