Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nextteq™ Gastec™ Detector Tubes: Carbon Dioxide
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
$69.24 - $201.50
Specifications
Components Measured | Carbon Dioxide |
---|---|
Quantity | 10 |
Temperature Range (English) | 32°F to 104°F |
Temperature Range (Metric) | 0°C to 40°C |
Catalog Number | Mfr. No. | Components Measured | Measurement Range | Response Time | No. of Strokes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Components Measured | Measurement Range | Response Time | No. of Strokes | Price | Quantity & Availability | ||
19-026-261
![]() |
Nextteq™
2L |
Carbon Dioxide | 0.13 to 6% | 45 sec. to 3 min. | 0.5 to 2 |
Pack of 10 for $201.50
|
|
||
19-026-263
![]() |
Nextteq™
2LL |
Carbon Dioxide | 300 to 5000 | 1.5 min. | 1 |
Pack of 10 for $69.24
Case of 10 PK for $1,181.00
|
|
||
Description
Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.
Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.Specifications
Carbon Dioxide | |
10 | |
32°F to 104°F | |
0°C to 40°C |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title