Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Newton's Second Law of Motion Apparatus
Demonstrate Newton's Second Law of Motion with a device that can be mounted to a support stand or a table clamp. The Newton's Second Law of Motion Apparatus is made of solid metal and designed to last many years in the classroom.
Supplier: Eisco™ PH0319
Description
- This Second Law of Motion Apparatus is made of solid metal and designed to last many years in the class room
- Base is painted blue and is 10 in. x 2.5 in.
- Spring driven and offers two tensions
- Includes 7.5 in. x 0.50 in. diameter rod to be able to mount on table clamp or fixed to a support stand
- Complete with two steel balls
Specifications
Newton's Law Demonstrator | |
2 Steel Balls, Rod (7.5 in. L x 0.5 in. dia.) | |
Blue |
Steel | |
30 x 12 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction