Learn More
R & D SYSTEMS INC Newborn Calf Serum
Supplier:  R & D SYSTEMS INC S11295
Encompass_Preferred
Call your Customer Service Representative for Bulk Discounts. R&D Systems Newborn Calf Serum (NBC, NBCS) provides many of the same nutrients as Fetal Bovine Serum, but has slightly higher protein levels. For routine maintenance of select cell lines and where low antibody concentrations are not critical, Newborn Calf Serum is often used as a lower-priced alternative to FBS. All R&D Systems Sera are manufactured in an ISO 9001:2015 certified facility.
Catalog No. 50-152-7074