Learn More
Eppendorf™ Scientific Shaker Utility Carriers
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Supplier:  Eppendorf™ M11909907
 View more versions of this product
Catalog No. 05-414-397
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • For shaking vessels without fasteners
  • Carriers have rubber-cushioned retainer bars
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Utility Carrier
30.5 cm
41 cm
Retainer bars
12 in.
16 in.
For Innova 2050 Benchtop open-air shaker
Product Suggestions
Product Suggestions