Learn More
Immy MycoDDR Neutralization Buffer B, Individual Bulk, Immy TB-PN67-60
Supplier: Immy TBPN678
MycoDDR Neutralization Buffer B is a reagent used in the MycoDDR TB Detection Kit by IMmy. It is sold individually or in bulk quantities. The buffer neutralizes the amplification reaction to prevent carryover contamination. It should be stored at 2-8°C upon receipt. This product allows detection of Mycobacterium tuberculosis complex and resistance to rifampin. When used with the kit, it enables a rapid, accurate diagnosis of TB and drug resistance.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.