Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Neuropathiazol
Selective inducer of neuronal differentiation in hippocampal neural progenitors
$185.60 - $770.24
Specifications
Purity | >98% |
---|---|
Content And Storage | -20°C |
Product Type | Neuropathiazol |
Molecular Weight (g/mol) | 338.42 |
Inhibitors | Neuropathiazol |
Description
Selective inducer of neuronal differentiation in cultured hippocampal neural progenitor cells (NPCs). Selectively suppresses astrocyte differentiation when induced by LIF, BMP2 and FBS compared to retinoic acid. Enhances maturation of NPC derived neurons.Specifications
>98% | |
Neuropathiazol | |
Neuropathiazol |
-20°C | |
338.42 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title