Learn More
STEMCELL Technologies NeuroCult™ Neuronal Plating Medium , Size: 100 mL
Supplier: STEMCELL Technologies 05713
NeuroCult™ Neuronal Plating Medium is a serum-free neuronal basal medium. It is designed for use with BrainPhys™ Neuronal Medium (Catalog #05790) for the plating and culture of primary tissue-derived neurons.