Learn More
VWR INTERNATIONAL LLC NESSLER COLOR COMPARISON TUBES
Supplier: VWR INTERNATIONAL LLC 66176507
KIMAX® Nessler Color Comparison Tubes, Tall Form, APHA Standard, Kimble Chase. Matched Set. Scale length: 200-220mm. Tube dimensions: 20 x 300mm. Graduation increments: 50 mL