Learn More
LABSYSTEMS DIAGNOSTICS OY Determiniation of GALT activity from DBS
Supplier: LABSYSTEMS DIAGNOSTICS OY 6199810
Neonatal GALT test is designed for the quantitative in vitro determination of galactose-1-phosphate uridyltransferase (GALT) activity in blood samples dried on filter paper intended for screening newborns for classic galactosemia caused by galactose-1-phosphate uridyltransferase deficiency. The kit can be used for both manual and automate use
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.