Learn More
Neogen Diagnostics Testosterone ELISA Kit - 96 well
Supplier: Neogen Diagnostics 402510
Testosterone ELISA is used for the quantitative analysis of Testosterone levels in urine, oral fluid, tissue culture supernatant, plasma or serum samples and is non-species specific. Assay range: 0.002 - 2 ng/mL, Assay sample size: 50 µL.