Learn More
Neogen Diagnostics Corticosterone ELISA Kit - 96 well
Supplier: Neogen Diagnostics 402810
Corticosterone ELISA is used for the quantitative analysis of Corticosterone levels in urine, oral fluid, tissue culture supernatant, plasma or serum samples and is non-species specific. Assay range: 0.05 - 5 ng/mL assay range, Assay sample size: 50 µL.