Learn More
Sklar Instruments Nelson Trocar, Sklar, 8-1/4 in., Stainless Steel, #3, 33 French. EA/1.
SDP
Supplier:  Sklar Instruments 342434
Encompass_Preferred
Nelson Trocar, Sklar, 8-1/4 in., Stainless Steel, #3, 33 French. Sklar's Nelson #3 Trocar is used for drainage fluids during cardiothoracic surgeries. The products are available in a variety of French sizesand has a bulb shaped hollow handle for a firm grip with a sharp tip for safe penetration.
Catalog No. 50-143-8250