Learn More
BTX™ Needles Array Electrode Handles for BTX™ AgilePulse™ in Vivo
Needle Array Handle for miniature needles designed to provide superior, highly-uniform electric fields in dermal or muscular tissue as part of the AgilePulse™ in vivo Gene Delivery Electroporation System. Specially designed for gene and vaccine delivery.
Supplier:  BTX™ 470092
 View more versions of this product
Catalog No. BTX470092
Add to Cart
Description
Description
The miniature parallel-needle array is inserted directly into the target site for fast, reliable in vivo electroporation. The electric fields produced are the closest approximation of parallel plate electrodes, treating approximately 80% of the target area with 95% of the applied electric field.
The needle array requires the needle array handle which includes the electrode connector cable. Several needle lengths and row spacing are available for various intra-dermal or intra-muscular applications. Needle Arrays can be used for up to 500 pulses with proper care and maintenance.
  • Uniform, reliable electric fields
  • Miniature needles minimize tissue trauma
  • Disposable and sterile
  • Medical-grade plastic and surgical steel construction
  • Safety-assured design
  • Multiple configurations, sizes to choose from
Specifications
Specifications
Gene and Vaccine Delivery
Plastic
Needle Array Handle
1 Ea.
Electrode Connector Cable
Generators with SN AP-4069-ID/AP-5031-IM and After
BTX Electroporation
Handle Only
Non-sterile
Square Wave
Product Suggestions
Product Suggestions