Learn More
Perkin Elmer US LLC Nebulizer Cleaning Wires, Pkg. 5
Supplier: Perkin Elmer US LLC 03030135
It is recommended to use only PerkinElmer cleaning wires to clean PerkinElmer nebulizers. They are included in the Stainless Steel Nebulizer Capillary Assembly for for AAnalyst Models 3100/3110/3300/5000/5100 PC (03030354) and the Corrosion-Resistant Nebulizer Complete Assembly for AAnalyst Models 3100/3110/3300/5000/5100 PC (03030404).