Learn More
New England Biolabs, Inc. NEBuffer Set (EcoRI, DpnII) – 1.25 ml of each
SDP
Supplier:  New England Biolabs, Inc. B7006S
Encompass
New England Biolabs provides 10X NEBuffer with each restriction endonuclease to assure optimal (100%) activity. Set contains two 1.25ml vials of each unique NEBuffer for the enzymes EcoRI and DpnII. Most of our enzymes are supplied with one of four standard NEBuffers. Occasionally, an enzyme has specific buffer requirements not met by one of the four standard NEBuffers, in which case the enzyme is supplied with its own unique NEBuffer. 1X NEBuffer EcoRI 100 mM Tris-HCl 50 mM NaCl 10 mM MgCl2 0.025% Triton X-100 (pH 7.5 @ 25C) 1X NEBuffer DpnII 50 mM Bis-Tris-HCl 100 mM NaCl 10 mM MgCl2 1 mM DTT (pH 6 @ 25C)
Catalog No. 50-152-7894