Learn More
New England Biolabs, Inc. NEBNext UltraExpress RNA 96
Supplier: New England Biolabs, Inc. E3330L
The NEBNext UltraExpress RNA Library Prep Kit is the latest generation of NEBNext RNA library prep, and has a fast, streamlined workflow. With a 3-hour library prep protocol, the kit enables creation of high-quality RNA libraries in a single day, in conjunction with mRNA enrichment or rRNA depletion kits.