Learn More
Zeptometrix NATtrol™ M. catarrhalis Stock (Quantitative)
Designed to evaluate the performance of nucleic acid tests for determination of the presence of M. catarrhalis nucleic acid.
Supplier: Zeptometrix NATMCASTQ
Description
NATtrol™ M. catarrhalis Stock is formulated with purified, intact bacterial cells that have been chemically modified to render them non-infectious and refrigerator stable. Each vial contains 1.0 mL of NATtrol™ M. catarrhalis stock in a proprietary matrix with a target concentration.
INTENDED USE:
- NATtrol™ M. catarrhalis stock is designed to evaluate the performance of nucleic acid tests for determination of the presence of M. catarrhalis nucleic acid.
- NATtrol™ M. catarrhalis stock enables laboratories to monitor test variation, lot-to-lot test kit performance, operator variation, and can provide assistance in identifying random or systemic error.
Specifications
ISO 13485 | |
NATtrol Virus Stock | |
1 mL |
1,000,000 cp/mL | |
Evaluate performance of molecular tests, verification of assays, training of laboratory personnel and to monitor assay-kit lot performance | |
RUO |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
RUO only.