Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Warning Biological Hazard Sign
Alerts employees and guests when a hazard is present in a facility
Specifications
Certifications/Compliance | ANSI |
---|---|
Quantity | 1 Ea. |
Height (English) | 10 in. |
Width (English) | 7 in. |
Displayed Text | Warning Biological Hazard |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
19-065-126
|
National Marker™
WGA5P |
Vinyl | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
19-065-125
|
National Marker™
WGA5R |
Plastic | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Orange, white and black sign with white and black text
- Available in rigid plastic or pressure-sensitive vinyl
- Size: 25.4 × 17.7cm (10 × 7 in.)
Specifications
ANSI | |
10 in. | |
Warning Biological Hazard |
1 Ea. | |
7 in. | |
Warning Sign |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title