Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Notice In Accordance with OSHA Regulations Signs
Supplier: National Marker™ N140RB
Specifications
Notice In Accordance With OSHA Regulations | |
Plastic | |
Rigid Plastic | |
0.05 in. | |
14 in. |
OSHA 29 CFR 1910.145 | |
1 Ea. | |
10 in. | |
Safety Sign |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title